3-Benzyloxyphenylacetonitril 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(3-(Benzyloxy)phenyl)acetonitrile

OR4462-1G OR4462-5G OR4462-25G
APOSOR4462-1GEA 436 DKK
APOSOR4462-1G APOSOR4462-5G APOSOR4462-25G
3-Benzyloxyphenylacetonitril 95%
3-Benzyloxyphenylacetonitril
Formel: C₁₅H₁₃NO
Kogepunkt: 188…190 °C/3mmHg
Smeltepunkt: 47…50 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016396
CAS-nummer: 20967-96-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR