2-Amino-4,6-dimethoxy-1,3,5-triazine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR4381-100G OR4381-25G OR4381-5G
APOSOR4381-100GEA 710 DKK
APOSOR4381-100G APOSOR4381-25G APOSOR4381-5G
2-Amino-4,6-dimethoxy-1,3,5-triazine 98%
2-Amino-4,6-dimethoxy-1,3,5-triazine
Formel: C₅H₈N₄O₂
Smeltepunkt: 220…222 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052765
CAS-nummer: 16370-63-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR