5-Bromo-2-chloroquinoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR43543-1G OR43543-250MG
APOSOR43543-1GEA 3360 DKK
APOSOR43543-1G APOSOR43543-250MG
5-Bromo-2-chloroquinoline
5-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
Molekylvægt: 242.4997 g/mol
Smeltepunkt: 71…78 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08437594
CAS-nummer: 99455-13-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR