3-Phenylpiperidin 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR43508-250MG OR43508-5G OR43508-1G
APOSOR43508-250MGEA 371 DKK
APOSOR43508-250MG APOSOR43508-5G APOSOR43508-1G
3-Phenylpiperidin 97%
3-Phenylpiperidin
Formel: C₁₁H₁₅N
Kogepunkt: 114 °C/8mmHg
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00804860
CAS-nummer: 3973-62-4
EINECS: 223-602-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR