6-Nitroquinaldin 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methyl-6-nitroquinoline

OR4322-25G OR4322-5G
APOSOR4322-25GEA 5060 DKK
APOSOR4322-25G APOSOR4322-5G
6-Nitroquinaldin 97%
6-Nitroquinaldin
Formel: C₁₀H₈N₂O₂
Molekylvægt: 188.19 g/mol
Smeltepunkt: 162…166 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051736
CAS-nummer: 613-30-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR