7-(Diethylamino)-4-methylcoumarin 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR42234-100G OR42234-500G
APOSOR42234-100GEA 186 DKK
APOSOR42234-100G APOSOR42234-500G
7-(Diethylamino)-4-methylcoumarin 98%
7-(Diethylamino)-4-methylcoumarin
Formel: C₁₄H₁₇NO₂
Kogepunkt: 240 °C/6,5mmHg
Smeltepunkt: 72…75 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006864
CAS-nummer: 91-44-1
EINECS: 202-068-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR