3-Chloro-4-methylpyridazine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR42178-1G OR42178-250MG
APOSOR42178-1GEA 1810 DKK
APOSOR42178-1G APOSOR42178-250MG
3-Chloro-4-methylpyridazine 95%
3-Chloro-4-methylpyridazine
Formel: C₅H₅ClN₂
Smeltepunkt: 46…47 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08460979
CAS-nummer: 68206-04-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR