(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR4201-5G OR4201-10G
APOSOR4201-5GEA 871 DKK
APOSOR4201-5G APOSOR4201-10G
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid
Formel: C₁₁H₁₂O₂
Smeltepunkt: 55…57 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07368365
CAS-nummer: 85977-52-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR