N-Benzyl-2-chloroacetamide
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR4110-1G
OR4110-5G
APOSOR4110-1GEA
436
DKK
APOSOR4110-1G
APOSOR4110-5G
N-Benzyl-2-chloroacetamide
N-Benzyl-2-chloroacetamide
Formel:
C₉H₁₀ClNO Smeltepunkt: 93…96 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00084929 CAS-nummer: 2564-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...