3-Amino-N,N-dimethylanilin 2 HCl
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: N1,N1-Dimethylbenzene-1,3-diamine dihydrochloride, N,N-Dimethyl-1,3-phenylenediamine dihydrochloride, N,N-Dimethyl-m-phenylenediamine dihydrochloride
OR40391-25G OR40391-5G
APOSOR40391-25GEA 1940 DKK
APOSOR40391-25G APOSOR40391-5G
3-Amino-N,N-dimethylanilin 2 HCl
3-Amino-N,N-dimethylanilin 2 HCl
Formel: C₈H₁₄Cl₂N₂
Molekylvægt: 209.12 g/mol
Smeltepunkt: 218
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012971
CAS-nummer: 3575-32-4