1-Pyreneboronic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR40378-5G OR40378-10G
APOSOR40378-5GEA 251 DKK
APOSOR40378-5G APOSOR40378-10G
1-Pyreneboronic acid 97%
1-Pyreneboronic acid
Formel: C₁₆H₁₁BO₂
Smeltepunkt: 247…251 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04974062
CAS-nummer: 164461-18-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR