Chloroformamidine hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR40351-5G OR40351-1G
APOSOR40351-5GEA 0 DKK
APOSOR40351-5G APOSOR40351-1G
Chloroformamidine hydrochloride
Chloroformamidine hydrochloride
Formel: CH₄Cl₂N₂
Smeltepunkt: 177 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00035527
CAS-nummer: 29671-92-9
UN nummer: 1759
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR