3-Amino-5-chloro-2-methoxyphenylbenzenecarboxylate 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Amino-5-chloro-2-methoxyphenyl benzoate

OR40341-1G OR40341-500MG
APOSOR40341-1GEA 7990 DKK
APOSOR40341-1G APOSOR40341-500MG
3-Amino-5-chloro-2-methoxyphenylbenzenecarboxylate 95+%
3-Amino-5-chloro-2-methoxyphenylbenzenecarboxylate
Formel: C₁₄H₁₂ClNO₃
Molekylvægt: 277.71 g/mol
Smeltepunkt: 144…145 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16622765
CAS-nummer: 1228182-63-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR