Methyl-2-(benzyloxy)-5-methoxy-3-nitrobenzenecarboxylate 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 2-(benzyloxy)-5-methoxy-3-nitrobenzoate

OR40312-1G OR40312-500MG OR40312-5G
APOSOR40312-1GEA 2030 DKK
APOSOR40312-1G APOSOR40312-500MG APOSOR40312-5G
Methyl-2-(benzyloxy)-5-methoxy-3-nitrobenzenecarboxylate 95+%
Methyl-2-(benzyloxy)-5-methoxy-3-nitrobenzenecarboxylate
Formel: C₁₆H₁₅NO₆
Smeltepunkt: 76…78 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16140356
CAS-nummer: 1221792-98-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR