1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
7-Azaindole-5-carboxylic acid
OR40241-1G
OR40241-5G
OR40241-10G
APOSOR40241-1GEA
305
DKK
APOSOR40241-1G
APOSOR40241-5G
APOSOR40241-10G
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid 97%
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid
Formel:
C₈H₆N₂O₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08272240 CAS-nummer: 754214-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...