2-Chloropyrimidine-5-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR40235-1G OR40235-5G
APOSOR40235-1GEA 164 DKK
APOSOR40235-1G APOSOR40235-5G
2-Chloropyrimidine-5-carboxylic acid
2-Chloropyrimidine-5-carboxylic acid
Formel: C₅H₃ClN₂O₂
Molekylvægt: 158.5400 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06739036
CAS-nummer: 374068-01-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR