3-(2-Thiazolylcarbamoyl)phenylboronic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR4022-1G OR4022-500MG
APOSOR4022-1GEA 1130 DKK
APOSOR4022-1G APOSOR4022-500MG
3-(2-Thiazolylcarbamoyl)phenylboronic acid 97%
3-(2-Thiazolylcarbamoyl)phenylboronic acid
Formel: C₁₀H₉BN₂O₃S
Smeltepunkt: 218…222 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06659885
CAS-nummer: 850567-34-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR