(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2R)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected, (R)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

OR40210-1G OR40210-5G
APOSOR40210-1GEA 622 DKK
APOSOR40210-1G APOSOR40210-5G
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid 95%
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid
Formel: C₁₁H₁₇NO₅
Smeltepunkt: 119…124 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09953681
CAS-nummer: 1212176-33-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR