5-Aminoquinolin 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Quinolin-5-amine, Quinolin-5-ylamine

OR4011-25G OR4011-5G
APOSOR4011-25GEA 393 DKK
APOSOR4011-25G APOSOR4011-5G
5-Aminoquinolin 97%
5-Aminoquinolin
Formel: C₉H₈N₂
Kogepunkt: 310 °C
Smeltepunkt: 107…109 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006797
CAS-nummer: 611-34-7
EINECS: 210-266-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR