2-Brom-6-methoxybenzoesyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR400083-1G OR400083-25G OR400083-5G
APOSOR400083-1GEA 183 DKK
APOSOR400083-1G APOSOR400083-25G APOSOR400083-5G
2-Brom-6-methoxybenzoesyre
2-Brom-6-methoxybenzoesyre
Formel: C₈H₇BrO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00800732
CAS-nummer: 31786-45-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR