Pyrimidine-4-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR3737-1G
OR3737-5G
OR3737-10G
APOSOR3737-1GEA
164
DKK
APOSOR3737-1G
APOSOR3737-5G
APOSOR3737-10G
Pyrimidine-4-carboxylic acid
Pyrimidine-4-carboxylic acid
Formel:
C5H4N2O2 Molekylvægt: 124.10 g/mol Smeltepunkt: 229.8…231.9 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00129737 CAS-nummer: 31462-59-6 EINECS: 250-641-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...