2-Aminooxazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Oxazol-2-amine
OR3721-1G
OR3721-5G
APOSOR3721-1GEA
175
DKK
APOSOR3721-1G
APOSOR3721-5G
2-Aminooxazole
2-Aminooxazole
Formel:
C₃H₄N₂O Molekylvægt: 84.10 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07364485 CAS-nummer: 4570-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...