5-Brom-2-chlor-3-nitropyridin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR3580-25G OR3580-100G OR3580-5G
APOSOR3580-25GEA 198 DKK
APOSOR3580-25G APOSOR3580-100G APOSOR3580-5G
5-Brom-2-chlor-3-nitropyridin
5-Brom-2-chlor-3-nitropyridin
Formel: C₅H₂BrClN₂O₂
Smeltepunkt: 65…70 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00222270
CAS-nummer: 67443-38-3
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR