2,5-Dibromopyrimidine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
OR350122-100G
OR350122-25G
APOSOR350122-100GEA
2810
DKK
APOSOR350122-100G
APOSOR350122-25G
2,5-Dibromopyrimidine
2,5-Dibromopyrimidine
Formel:
C₄H₂Br₂N₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08275684 CAS-nummer: 32779-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...