4-(Benzyloxy)benzenesulphonyl chloride 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR350065-1G OR350065-5G OR350065-250MG OR350065-500MG
APOSOR350065-1GEA 162 DKK
APOSOR350065-1G APOSOR350065-5G APOSOR350065-250MG APOSOR350065-500MG
4-(Benzyloxy)benzenesulphonyl chloride 98%
4-(Benzyloxy)benzenesulphonyl chloride
Formel: C₁₃H₁₁ClO₃S
Smeltepunkt: 102…105 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04035557
CAS-nummer: 87001-32-9
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR