2-Chloropyrimidine-5-sulphonyl chloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR346309-1G OR346309-5G
APOSOR346309-1GEA 959 DKK
APOSOR346309-1G APOSOR346309-5G
2-Chloropyrimidine-5-sulphonyl chloride
2-Chloropyrimidine-5-sulphonyl chloride
Formel: C₄H₂Cl₂N₂O₂S
Smeltepunkt: 64…67 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06657208
CAS-nummer: 98026-88-1
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR