5-(2-Thienyl)isoxazole-3-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-(Thiophen-2-yl)isoxazole-3-carboxylic acid

OR346206-1G OR346206-5G
APOSOR346206-1GEA 687 DKK
APOSOR346206-1G APOSOR346206-5G
5-(2-Thienyl)isoxazole-3-carboxylic acid
5-(2-Thienyl)isoxazole-3-carboxylic acid
Formel: C₈H₅NO₃S
Molekylvægt: 195.192 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04113955
CAS-nummer: 763109-71-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR