3-Chloro-4-nitropyridine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR345250-10G
OR345250-1G
OR345250-5G
APOSOR345250-10GEA
2710
DKK
APOSOR345250-10G
APOSOR345250-1G
APOSOR345250-5G
3-Chloro-4-nitropyridine
3-Chloro-4-nitropyridine
Formel:
C5H3ClN2O2 Molekylvægt: 158.54 g/mol Kogepunkt: 256 °C Smeltepunkt: 25…26 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00490146 CAS-nummer: 13194-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...