Methyl amino(4-methylphenyl)acetate hydrochloride 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 2-amino-2-(p-tolyl)acetate hydrochloride

OR33515-1G OR33515-5G
APOSOR33515-1GEA 2520 DKK
APOSOR33515-1G APOSOR33515-5G
Methyl amino(4-methylphenyl)acetate hydrochloride 95%
Methyl amino(4-methylphenyl)acetate hydrochloride
Formel: C₁₀H₁₄ClNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08456676
CAS-nummer: 134722-09-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR