Methyl2-chlornicotinat 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR3350-100G
OR3350-25G
OR3350-500G
APOSOR3350-100GEA
218
DKK
APOSOR3350-100G
APOSOR3350-25G
APOSOR3350-500G
Methyl2-chlornicotinat 98%
Methyl2-chlornicotinat
Formel:
C₇H₆ClNO₂ Kogepunkt: 150 °C/1mmHg Massefylde: 1,314 g/cm³ (25 °C) Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01318629 CAS-nummer: 40134-18-7 EINECS: 254-805-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...