2-Chloro-5-methoxyphenol 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
OR3294-25G
OR3294-5G
OR3294-100G
APOSOR3294-25GEA
1830
DKK
APOSOR3294-25G
APOSOR3294-5G
APOSOR3294-100G
2-Chloro-5-methoxyphenol 95%
2-Chloro-5-methoxyphenol
Formel:
C₇H₇ClO₂ Kogepunkt: 125 °C/10mmHg Massefylde: 1,31 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00086345 CAS-nummer: 18113-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...