(R)-(+)-β-Methylphenethylamine 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(R)-(+)-beta-Methylphenethylamine, (R)-(+)-2-Phenyl-1-propylamine
OR3244-25G
OR3244-5G
OR3244-100G
APOSOR3244-25GEA
1640
DKK
APOSOR3244-25G
APOSOR3244-5G
APOSOR3244-100G
(R)-(+)-β-Methylphenethylamine 98%
(R)-(+)-β-Methylphenethylamine
Formel:
C₉H₁₃N Kogepunkt: 197 °C Massefylde: 0.945 g/cm³ (25 °C) Flammepunkt: 81 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00216740 CAS-nummer: 28163-64-6 UN nummer: 2735 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...