2-Methyl-2-piperidinopropanal 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR32364-1G OR32364-5G
APOSOR32364-1GEA 2030 DKK
APOSOR32364-1G APOSOR32364-5G
2-Methyl-2-piperidinopropanal 95%
2-Methyl-2-piperidinopropanal
Formel: C₉H₁₇NO
Molekylvægt: 155.24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08056609
CAS-nummer: 16042-93-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR