Boc-4-phenylpiperidine-4-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR322299-1G
OR322299-250MG
APOSOR322299-1GEA
441
DKK
APOSOR322299-1G
APOSOR322299-250MG
Boc-4-phenylpiperidine-4-carboxylic acid
Boc-4-phenylpiperidine-4-carboxylic acid
Formel:
C₁₇H₂₃NO₄ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00673774 CAS-nummer: 167262-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...