4-Chloro-2-(pyridin-2-yl)-6-(pyridin-3-yl)pyrimidine 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR318039-1G OR318039-5G OR318039-10G
APOSOR318039-1GEA 4000 DKK
APOSOR318039-1G APOSOR318039-5G APOSOR318039-10G
4-Chloro-2-(pyridin-2-yl)-6-(pyridin-3-yl)pyrimidine 95+%
4-Chloro-2-(pyridin-2-yl)-6-(pyridin-3-yl)pyrimidine
Formel: C₁₄H₉ClN₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10688276
CAS-nummer: 1001915-28-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR