2-Methyladamantan-2-yl acrylate 99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Acrylic acid 2-Methyladamantan-2-yl ester

OR315775-1G OR315775-5G OR315775-25G
APOSOR315775-1GEA 164 DKK
APOSOR315775-1G APOSOR315775-5G APOSOR315775-25G
2-Methyladamantan-2-yl acrylate 99%
2-Methyladamantan-2-yl acrylate
Formel: C₁₄H₂₀O₂
Molekylvægt: 220.31 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08704223
CAS-nummer: 249562-06-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR