4,6-Dibromodibenzothiophene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR315130-1G OR315130-250MG OR315130-5G OR315130-25G
APOSOR315130-1GEA 173 DKK
APOSOR315130-1G APOSOR315130-250MG APOSOR315130-5G APOSOR315130-25G
4,6-Dibromodibenzothiophene
4,6-Dibromodibenzothiophene
Formel: C₁₂H₆Br₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18451473
CAS-nummer: 669773-34-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR