3-Aminopyridin-4-carbonitril 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Amino-4-cyanopyridine

OR30946-1G OR30946-250MG OR30946-5G
APOSOR30946-1GEA 545 DKK
APOSOR30946-1G APOSOR30946-250MG APOSOR30946-5G
3-Aminopyridin-4-carbonitril 96%
3-Aminopyridin-4-carbonitril
Formel: C₆H₅N₃
Smeltepunkt: 135…138 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04035596
CAS-nummer: 78790-79-1
UN nummer: 3439
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR