3-Methyloxetan-3-ol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR309102-1G OR309102-5G
APOSOR309102-1GEA 1030 DKK
APOSOR309102-1G APOSOR309102-5G
3-Methyloxetan-3-ol 97%
3-Methyloxetan-3-ol
Formel: C₄H₈O₂
Opbevaringstemperatur: Fryser
MDL Number: MFCD18909291
CAS-nummer: 162816-08-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR