2-(2-Pyridyl)benzimidazole 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR30756-1G
OR30756-5G
OR30756-25G
APOSOR30756-1GEA
162
DKK
APOSOR30756-1G
APOSOR30756-5G
APOSOR30756-25G
2-(2-Pyridyl)benzimidazole 97%
2-(2-Pyridyl)benzimidazole
Formel:
C₁₂H₉N₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005586 CAS-nummer: 1137-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...