N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,4-butanediamine, NH2-C4-NH-Boc, N-Boc-1,4-diaminobutane, N-Boc-1,4-butanediamine, tert-Butyl N-(4-Aminobutyl)carbamate, tert-Butyl (4-aminobutyl)carbamate
OR307039-25G
OR307039-5G
OR307039-100G
APOSOR307039-25GEA
567
DKK
APOSOR307039-25G
APOSOR307039-5G
APOSOR307039-100G
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97%
N-(tert-Butoxycarbonyl)-1,4-diaminobutane
Formel:
C₉H₂₀N₂O₂ Massefylde: 0,984 g/cm³ (25 °C) Flammepunkt: 109 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00210019 CAS-nummer: 68076-36-8 UN nummer: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...