3-Methyl-4-nitrobenzaldehyde 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR30523-1G OR30523-5G
APOSOR30523-1GEA 262 DKK
APOSOR30523-1G APOSOR30523-5G
3-Methyl-4-nitrobenzaldehyde 97%
3-Methyl-4-nitrobenzaldehyde
Formel: C8H7NO3
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08445621
CAS-nummer: 18515-67-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR