2-Brom-4-methylbenzoesyre 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR3052-100G OR3052-25G
APOSOR3052-100GEA 1230 DKK
APOSOR3052-100G APOSOR3052-25G
2-Brom-4-methylbenzoesyre 97%
2-Brom-4-methylbenzoesyre
Formel: C₈H₇BrO₂
Smeltepunkt: 143…147 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00045797
CAS-nummer: 7697-27-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR