2-Brom-4-methylanisol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Bromo-2-methoxy-5-methylbenzene, 3-Bromo-4-methoxytoluene
OR3050-50G
OR3050-10G
OR3050-250G
APOSOR3050-50GEA
273
DKK
APOSOR3050-50G
APOSOR3050-10G
APOSOR3050-250G
2-Brom-4-methylanisol 98%
2-Brom-4-methylanisol
Formel:
C₈H₉BrO Kogepunkt: 134 °C/25mmHg Massefylde: 1,41 g/cm³ (25 °C) Flammepunkt: 113 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00017783 CAS-nummer: 22002-45-5 EINECS: 244-705-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...