4-Chloro-2-iodophenol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR30481-5G
OR30481-10G
OR30481-1G
APOSOR30481-5GEA
218
DKK
APOSOR30481-5G
APOSOR30481-10G
APOSOR30481-1G
4-Chloro-2-iodophenol 98%
4-Chloro-2-iodophenol
Formel:
C₆H₄ClIO Molekylvægt: 254.4500 g/mol Smeltepunkt: 74…76 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08235248 CAS-nummer: 71643-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...