Methyl-6-chloropyridazine-4-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR304309-1G OR304309-25G OR304309-5G
APOSOR304309-1GEA 173 DKK
APOSOR304309-1G APOSOR304309-25G APOSOR304309-5G
Methyl-6-chloropyridazine-4-carboxylate
Methyl-6-chloropyridazine-4-carboxylate
Formel: C₆H₅ClN₂O₂
Smeltepunkt: 88…89 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16660968
CAS-nummer: 1093860-48-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR