4-Chloro-2,6-pyridinedicarboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR303669-1G OR303669-250MG OR303669-5G
APOSOR303669-1GEA 183 DKK
APOSOR303669-1G APOSOR303669-250MG APOSOR303669-5G
4-Chloro-2,6-pyridinedicarboxylic acid
4-Chloro-2,6-pyridinedicarboxylic acid
Formel: C₇H₄ClNO₄
Smeltepunkt: 210 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04117894
CAS-nummer: 4722-94-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR