4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine, 4-(4-Methylpiperazino)methylphenylboronic acid, pinacol ester
OR303410-5G
OR303410-25G
APOSOR303410-5GEA
632
DKK
APOSOR303410-5G
APOSOR303410-25G
4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Formel:
C₁₈H₂₉BN₂O₂ Smeltepunkt: 51…55 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09266182 CAS-nummer: 938043-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...