O-Benzylhydroxylamine hydrochloride 97%
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
OR30224-100G OR30224-25G OR30224-5G
APOSOR30224-100GEA 535 DKK
APOSOR30224-100G APOSOR30224-25G APOSOR30224-5G
O-Benzylhydroxylamine hydrochloride 97%
O-Benzylhydroxylamine hydrochloride
Formel: C₇H₁₀ClNO
Molekylvægt: 159.62 g/mol
Smeltepunkt: 234…238 °C (subl.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012952
CAS-nummer: 2687-43-6
EINECS: 220-249-0