2-Amidinopyrimidine hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR301376-1G OR301376-5G
APOSOR301376-1GEA 183 DKK
APOSOR301376-1G APOSOR301376-5G
2-Amidinopyrimidine hydrochloride
2-Amidinopyrimidine hydrochloride
Formel: C₅H₇ClN₄
Smeltepunkt: 184…186 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02677965
CAS-nummer: 138588-40-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR